Product Details
- SNP ID
-
rs199889857
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99311579 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTAGCAGCATAGGGTTTATTGA[C/G]AGGTTCTGGGAAGCAGGAGGTACCC
- Phenotype
-
MIM: 103720
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADH1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13133908] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADH1B
- Gene Name
- alcohol dehydrogenase 1B (class I), beta polypeptide
View Full Product Details