Product Details

SNP ID
rs202102103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176215630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACTGTAAGAAATTCTTCAGTAA[C/T]GTTGGCAGCTGGAGTTGTGGGATAA
Phenotype
MIM: 615050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB5 PubMed Links

Gene Details

Gene
ASB5
Gene Name
ankyrin repeat and SOCS box containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080874.3 1765 Silent Mutation NP_543150.1
XM_005262759.1 1765 Silent Mutation XP_005262816.1
XM_011531616.2 1765 Silent Mutation XP_011529918.1
XM_011531617.2 1765 Silent Mutation XP_011529919.1

View Full Product Details