Product Details

SNP ID
rs202067618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154234564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGAACTTCATCTTCTGCTTTA[A/T]GTTCATGGCATGTACCACTGATGTG
Phenotype
MIM: 612486
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCHS2 PubMed Links

Gene Details

Gene
DCHS2
Gene Name
dachsous cadherin-related 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142552.1 6017 Intron NP_001136024.1
NM_017639.3 6017 Missense Mutation CAT,CTT H2908L NP_060109.2
XM_011532045.1 6017 Missense Mutation CAT,CTT H3363L XP_011530347.1
XM_011532046.2 6017 Missense Mutation CAT,CTT H2586L XP_011530348.1
XM_011532049.2 6017 Missense Mutation CAT,CTT H1698L XP_011530351.1
XM_017008320.1 6017 Intron XP_016863809.1

View Full Product Details