Product Details

SNP ID
rs201671383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:93829573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAACGCCTTGTCCGAGCTGCTAC[A/G]AACGCCCAGCGGAGGGGAACAGCCA
Phenotype
MIM: 601461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATOH1 PubMed Links

Gene Details

Gene
ATOH1
Gene Name
atonal bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005172.1 647 Missense Mutation CAA,CGA Q216R NP_005163.1

View Full Product Details