Product Details

SNP ID
rs201608727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1349724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACAGACCCCGCACAGCCTCTGC[C/T]GCCCCCCAGGGAGGCGGCACAGAGG
Phenotype
MIM: 606801 MIM: 614632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAEA PubMed Links

Gene Details

Gene
MAEA
Gene Name
macrophage erythroblast attacher
There are no transcripts associated with this gene.

Gene
UVSSA
Gene Name
UV stimulated scaffold protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317934.1 1069 Missense Mutation CCG,CTG P100L NP_001304863.1
NM_001317935.1 1069 Missense Mutation CCG,CTG P100L NP_001304864.1
NM_020894.3 1069 Missense Mutation CCG,CTG P100L NP_065945.2
XM_017008490.1 1069 Missense Mutation CCG,CTG P100L XP_016863979.1
XM_017008491.1 1069 Missense Mutation CCG,CTG P100L XP_016863980.1
XM_017008492.1 1069 Missense Mutation CCG,CTG P100L XP_016863981.1
XM_017008493.1 1069 Missense Mutation CCG,CTG P100L XP_016863982.1
XM_017008494.1 1069 Missense Mutation CCG,CTG P100L XP_016863983.1
XM_017008495.1 1069 Intron XP_016863984.1
XM_017008496.1 1069 Missense Mutation CCG,CTG P209L XP_016863985.1
XM_017008497.1 1069 Missense Mutation CCG,CTG P209L XP_016863986.1
XM_017008498.1 1069 Missense Mutation CCG,CTG P100L XP_016863987.1
XM_017008499.1 1069 Missense Mutation CCG,CTG P100L XP_016863988.1
XM_017008500.1 1069 Intron XP_016863989.1

View Full Product Details