Product Details
- SNP ID
-
rs201728277
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:103589779 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCGAGAGCAGCCATTGAAACTT[G/T]GGTCTCTTGGCGTTGCTCTTTTCTT
- Phenotype
-
MIM: 162332
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TACR3
PubMed Links
Gene Details
- Gene
- TACR3
- Gene Name
- tachykinin receptor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001059.2 |
1442 |
Missense Mutation |
|
|
NP_001050.1 |
View Full Product Details