Product Details

SNP ID
rs201335361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41982537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGGCTGTCGCTCCGGGGAGATC[C/T]TTGCCATTGATCTGCGTTGTAGAAA
Phenotype
MIM: 604604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF4L1 PubMed Links

Gene Details

Gene
DCAF4L1
Gene Name
DDB1 and CUL4 associated factor 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029955.3 Intron NP_001025126.2
Gene
SLC30A9
Gene Name
solute carrier family 30 member 9
There are no transcripts associated with this gene.

View Full Product Details