Product Details

SNP ID
rs202007257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151277184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGACAGAGACATGGGCCCTGC[A/T]GGCTGTGCCTTCACGCTGCTCCTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRSS48 PubMed Links

Gene Details

Gene
PRSS48
Gene Name
protease, serine 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183375.2 12 Silent Mutation GCA,GCT A4A NP_899231.2
XM_011531921.2 12 Intron XP_011530223.1
XM_011531922.2 12 Silent Mutation GCA,GCT A4A XP_011530224.1

View Full Product Details