Product Details
- SNP ID
-
rs202220769
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141421348 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGATATTCGGTGCCAGAAGAGACC[C/G]AAAAGGGCTCCTTCGTGGGCAATAT
- Phenotype
-
MIM: 606288
MIM: 606297
MIM: 606298
MIM: 603059
MIM: 606289
MIM: 606290
MIM: 606291
MIM: 606292
MIM: 606293
MIM: 606294
MIM: 606295
MIM: 606296
MIM: 606299
MIM: 606300
MIM: 606301
MIM: 603058
MIM: 606302
MIM: 606303
MIM: 606304
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHGA1
PubMed Links
Gene Details
- Gene
- PCDHGA1
- Gene Name
- protocadherin gamma subfamily A, 1
- Gene
- PCDHGA10
- Gene Name
- protocadherin gamma subfamily A, 10
- Gene
- PCDHGA11
- Gene Name
- protocadherin gamma subfamily A, 11
- Gene
- PCDHGA12
- Gene Name
- protocadherin gamma subfamily A, 12
There are no transcripts associated with this gene.
- Gene
- PCDHGA2
- Gene Name
- protocadherin gamma subfamily A, 2
- Gene
- PCDHGA3
- Gene Name
- protocadherin gamma subfamily A, 3
- Gene
- PCDHGA4
- Gene Name
- protocadherin gamma subfamily A, 4
- Gene
- PCDHGA5
- Gene Name
- protocadherin gamma subfamily A, 5
- Gene
- PCDHGA6
- Gene Name
- protocadherin gamma subfamily A, 6
- Gene
- PCDHGA7
- Gene Name
- protocadherin gamma subfamily A, 7
- Gene
- PCDHGA8
- Gene Name
- protocadherin gamma subfamily A, 8
- Gene
- PCDHGA9
- Gene Name
- protocadherin gamma subfamily A, 9
- Gene
- PCDHGB1
- Gene Name
- protocadherin gamma subfamily B, 1
- Gene
- PCDHGB2
- Gene Name
- protocadherin gamma subfamily B, 2
- Gene
- PCDHGB3
- Gene Name
- protocadherin gamma subfamily B, 3
- Gene
- PCDHGB4
- Gene Name
- protocadherin gamma subfamily B, 4
- Gene
- PCDHGB5
- Gene Name
- protocadherin gamma subfamily B, 5
- Gene
- PCDHGB6
- Gene Name
- protocadherin gamma subfamily B, 6
- Gene
- PCDHGB7
- Gene Name
- protocadherin gamma subfamily B, 7
- Gene
- PCDHGB8P
- Gene Name
- protocadherin gamma subfamily B, 8 pseudogene
There are no transcripts associated with this gene.
View Full Product Details