Product Details

SNP ID
rs201955015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:41929768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTATGTATGGAGCTGTCACTT[A/C]TTTTTTACACTCCCTGATCATTCAG
Phenotype
MIM: 609090
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C5orf51 PubMed Links

Gene Details

Gene
C5orf51
Gene Name
chromosome 5 open reading frame 51
There are no transcripts associated with this gene.

Gene
FBXO4
Gene Name
F-box protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297437.1 558 Missense Mutation TAT,TCT Y166S NP_001284366.1
NM_012176.2 558 Missense Mutation TAT,TCT Y166S NP_036308.1
NM_033484.2 558 Missense Mutation TAT,TCT Y166S NP_277019.1
XM_006714470.3 558 Missense Mutation TAT,TCT Y22S XP_006714533.1
XM_011514026.2 558 Missense Mutation TAT,TCT Y166S XP_011512328.1
XM_011514027.2 558 Missense Mutation TAT,TCT Y32S XP_011512329.1

View Full Product Details