Product Details
- SNP ID
-
rs202055986
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:77630511 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCCCATGGAGTTGGGCGGCGGA[A/C]CGGCCGCCAGGCTGCACTGGGACAG
- Phenotype
-
MIM: 604529
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OTP
PubMed Links
Gene Details
- Gene
- OTP
- Gene Name
- orthopedia homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032109.2 |
879 |
Missense Mutation |
GGT,GTT |
G244V |
NP_115485.1 |
- Gene
- WDR41
- Gene Name
- WD repeat domain 41
There are no transcripts associated with this gene.
View Full Product Details