Product Details

SNP ID
rs201939948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34922767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGCCCCTTTTGTCTTTTGACC[C/G]TGTAAGTTTCTCATTCAGTGTATGA
Phenotype
MIM: 603153
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRIX1 PubMed Links

Gene Details

Gene
BRIX1
Gene Name
BRX1, biogenesis of ribosomes
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018321.3 533 Missense Mutation CCT,CGT P170R NP_060791.3
Gene
DNAJC21
Gene Name
DnaJ heat shock protein family (Hsp40) member C21
There are no transcripts associated with this gene.

Gene
RAD1
Gene Name
RAD1 checkpoint DNA exonuclease
There are no transcripts associated with this gene.

View Full Product Details