Product Details

SNP ID
rs200196384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141924609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCGTACACTGGGACCTAGCCT[C/G]TGGAGGGTGACTCCTAAGTCCACCA
Phenotype
MIM: 615741
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0141 PubMed Links

Gene Details

Gene
KIAA0141
Gene Name
KIAA0141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142603.2 217 Silent Mutation CTC,CTG L20L NP_001136075.1
NM_014773.4 217 Silent Mutation CTC,CTG L20L NP_055588.3
XM_005268547.3 217 Silent Mutation CTC,CTG L20L XP_005268604.1
XM_005268548.3 217 Silent Mutation CTC,CTG L20L XP_005268605.1
XM_005268552.2 217 Silent Mutation CTC,CTG L20L XP_005268609.1
XM_006714812.3 217 Intron XP_006714875.1
XM_011537715.1 217 Silent Mutation CTC,CTG L20L XP_011536017.1
XM_011537716.2 217 Silent Mutation CTC,CTG L20L XP_011536018.1

View Full Product Details