Product Details

SNP ID
rs199762939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:124639189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTGGGCAGCCACCTGCTGAGA[G/T]GCAACAAGGGCAGCAGAGGTCAAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF608 PubMed Links

Gene Details

Gene
ZNF608
Gene Name
zinc finger protein 608
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020747.2 3847 Silent Mutation GCA,GCC A1492A NP_065798.2
XM_005272037.2 3847 Silent Mutation GCA,GCC A1492A XP_005272094.1
XM_005272038.1 3847 Silent Mutation GCA,GCC A1492A XP_005272095.1
XM_005272039.2 3847 Silent Mutation GCA,GCC A1492A XP_005272096.1
XM_005272040.3 3847 Silent Mutation GCA,GCC A1204A XP_005272097.1
XM_011543520.2 3847 UTR 3 XP_011541822.1
XM_011543522.2 3847 Silent Mutation GCA,GCC A1195A XP_011541824.1
XM_011543523.2 3847 Silent Mutation GCA,GCC A1107A XP_011541825.1
XM_017009674.1 3847 Silent Mutation GCA,GCC A1492A XP_016865163.1

View Full Product Details