Product Details

SNP ID
rs200121616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151746430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGCAGACCTTCTTGTTGGGCAG[G/T]TCAATGTCATACTTAACTCCTGCAG
Phenotype
MIM: 602270
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATOX1 PubMed Links

Gene Details

Gene
ATOX1
Gene Name
antioxidant 1 copper chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004045.3 200 Missense Mutation GAA,GAC E34D NP_004036.1

View Full Product Details