Product Details

SNP ID
rs200259581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176597844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATCCACCTTCTCTGAGGCCTGT[C/G]GTGTCTCCTTTTTCAAACACACAGC
Phenotype
MIM: 611239
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDHR2 PubMed Links

Gene Details

Gene
CDHR2
Gene Name
cadherin related family member 2
There are no transcripts associated with this gene.

Gene
GPRIN1
Gene Name
G protein regulated inducer of neurite outgrowth 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052899.2 2166 Missense Mutation CCA,CGA P664R NP_443131.2

View Full Product Details