Product Details

SNP ID
rs200322626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111071922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTGCCTTACTATATGTTCTGT[C/T]AGTTTCTTTCAGGAAAATCTTCATC
Phenotype
MIM: 607003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSLP PubMed Links

Gene Details

Gene
TSLP
Gene Name
thymic stromal lymphopoietin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033035.4 231 Missense Mutation TCA,TTA S11L NP_149024.1
NM_138551.4 231 Intron NP_612561.2
XM_011543698.1 231 Intron XP_011542000.1

View Full Product Details