Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018900.3 | 295 | Intron | NP_061723.1 | ||
NM_031410.2 | 295 | Intron | NP_113598.1 | ||
NM_031411.2 | 295 | Intron | NP_113599.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018901.3 | 295 | Missense Mutation | TTC,TTG | F43L | NP_061724.1 |
NM_031859.2 | 295 | Missense Mutation | TTC,TTG | F43L | NP_114065.1 |
NM_031860.2 | 295 | Missense Mutation | TTC,TTG | F43L | NP_114066.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018905.2 | 295 | Intron | NP_061728.1 | ||
NM_031495.1 | 295 | Intron | NP_113683.1 | ||
NM_031496.1 | 295 | Intron | NP_113684.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018906.2 | 295 | Intron | NP_061729.1 | ||
NM_031497.1 | 295 | Intron | NP_113685.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018907.3 | 295 | Intron | NP_061730.1 | ||
NM_031500.2 | 295 | Intron | NP_113688.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018908.2 | 295 | Intron | NP_061731.1 | ||
NM_031501.1 | 295 | Intron | NP_113689.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018909.3 | 295 | Intron | NP_061732.1 | ||
NM_031848.2 | 295 | Intron | NP_114036.1 | ||
NM_031849.2 | 295 | Intron | NP_114037.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018910.2 | 295 | Intron | NP_061733.1 | ||
NM_031852.1 | 295 | Intron | NP_114040.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018911.2 | 295 | Intron | NP_061734.1 | ||
NM_031856.1 | 295 | Intron | NP_114062.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014005.4 | 295 | Intron | NP_054724.1 | ||
NM_031857.1 | 295 | Intron | NP_114063.1 |