Product Details

SNP ID
rs199514346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1320623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACGCAGCCGCACTCACCACCGCT[A/G]GTACAGGTAGACCAGAAACACCACG
Phenotype
MIM: 612585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPTM1L PubMed Links

Gene Details

Gene
CLPTM1L
Gene Name
CLPTM1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030782.4 1718 Nonsense Mutation CAG,TAG Q509* NP_110409.2
XM_011514144.2 1718 Nonsense Mutation CAG,TAG Q508* XP_011512446.1

View Full Product Details