Product Details

SNP ID
rs199651608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141654055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGGGGCTGGGGGATTGGGGGCT[A/G]GATGGCTGGGAAGGACTTCCCAGGC
Phenotype
MIM: 606647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARAP3 PubMed Links

Gene Details

Gene
ARAP3
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022481.5 4027 Silent Mutation TCC,TCT S1510S NP_071926.4
XM_005268497.1 4027 Silent Mutation TCC,TCT S1497S XP_005268554.1
XM_005268498.1 4027 Silent Mutation TCC,TCT S1490S XP_005268555.1
XM_005268499.1 4027 Silent Mutation TCC,TCT S1432S XP_005268556.1
XM_005268500.3 4027 Intron XP_005268557.1
XM_006714792.1 4027 Silent Mutation TCC,TCT S1439S XP_006714855.1
XM_011537677.2 4027 Silent Mutation TCC,TCT S1315S XP_011535979.1
Gene
FCHSD1
Gene Name
FCH and double SH3 domains 1
There are no transcripts associated with this gene.

View Full Product Details