Product Details

SNP ID
rs199593296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56911253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGAACATTACTGAAAGTTTTCCA[A/G]GCTCTATTCTTAAATGATTTCAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIER3 PubMed Links

Gene Details

Gene
MIER3
Gene Name
MIER family member 3
There are no transcripts associated with this gene.

Gene
SETD9
Gene Name
SET domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171990.2 583 Silent Mutation CAA,CAG Q61Q NP_001165461.1
NM_001323018.1 583 Silent Mutation CAA,CAG Q35Q NP_001309947.1
NM_001323019.1 583 Silent Mutation CAA,CAG Q61Q NP_001309948.1
NM_001323020.1 583 Silent Mutation CAA,CAG Q35Q NP_001309949.1
NM_001323022.1 583 Intron NP_001309951.1
NM_153706.3 583 Silent Mutation CAA,CAG Q61Q NP_714917.2
XM_005248426.4 583 Intron XP_005248483.1
XM_011543139.2 583 Silent Mutation CAA,CAG Q29Q XP_011541441.1
XM_011543140.2 583 UTR 5 XP_011541442.1

View Full Product Details