Product Details
- SNP ID
-
rs201472333
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:181045533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCAGACTCCTTGAAGCCAGTAT[C/T]GCTGACCAGCAGTCTTGTCTTCCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BTNL9
PubMed Links
Gene Details
- Gene
- BTNL9
- Gene Name
- butyrophilin like 9
- Gene
- MIR8089
- Gene Name
- microRNA 8089
There are no transcripts associated with this gene.
View Full Product Details