Product Details

SNP ID
rs200819255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135936729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTTCTTCTAGACTGGGGGAGTTT[G/T]CCTCACCATGTAGTATTACAAATTT
Phenotype
MIM: 609087
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXL21 PubMed Links

Gene Details

Gene
FBXL21
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012159.4 617 Missense Mutation TTG,TTT L45F NP_036291.2

View Full Product Details