Product Details

SNP ID
rs201138474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150446906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAACATAGCAGCATATGGTGA[A/G]GATTCATCTCGGTCTGCCTTTACCT
Phenotype
MIM: 130620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS14 PubMed Links

Gene Details

Gene
RPS14
Gene Name
ribosomal protein S14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025070.1 480 Silent Mutation TCC,TCT S69S NP_001020241.1
NM_001025071.1 480 Silent Mutation TCC,TCT S69S NP_001020242.1
NM_005617.3 480 Intron NP_005608.1
XM_017009684.1 480 Silent Mutation TCC,TCT S69S XP_016865173.1

View Full Product Details