Product Details

SNP ID
rs200802497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170378935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTTCTCCACGTCGGCCCGGGC[C/T]GTCTGGTAATTGTCCACGCTGCCTG
Phenotype
MIM: 604660 MIM: 603951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNIP1 PubMed Links

Gene Details

Gene
KCNIP1
Gene Name
potassium voltage-gated channel interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034837.2 788 Intron NP_001030009.1
NM_001034838.2 788 Intron NP_001030010.1
NM_001278339.1 788 Intron NP_001265268.1
NM_001278340.1 788 Intron NP_001265269.1
NM_014592.3 788 Intron NP_055407.1
XM_017009407.1 788 Intron XP_016864896.1
XM_017009408.1 788 Intron XP_016864897.1
Gene
KCNMB1
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004137.3 788 Silent Mutation ACA,ACG T115T NP_004128.1

View Full Product Details