Product Details
- SNP ID
-
rs201146172
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:126603658 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTATCACATTATCGAGCTGTTGAAA[A/G]TGTGGATTCAAGTGAAGAAAGTTTT
- Phenotype
-
MIM: 107323
MIM: 604924
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH7A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3734173] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALDH7A1
- Gene Name
- aldehyde dehydrogenase 7 family member A1
There are no transcripts associated with this gene.
- Gene
- PHAX
- Gene Name
- phosphorylated adaptor for RNA export
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032177.3 |
|
Intron |
|
|
NP_115553.2 |
View Full Product Details