Product Details
- SNP ID
-
rs201323177
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:69195777 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAAGTTGCTTTTGAGATAAAAAA[G/T]CTTGCATTAGACAGGTAATTATTAC
- Phenotype
-
MIM: 605607
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CENPH
PubMed Links
Gene Details
- Gene
- CENPH
- Gene Name
- centromere protein H
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022909.3 |
387 |
Silent Mutation |
AAG,AAT |
K100N |
NP_075060.1 |
View Full Product Details