Product Details

SNP ID
rs201502793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:87268613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCCCTATCCTGGGCTGGTGGA[C/G]ACCGGAGTGGCTGGAACTCTGGGTG
Phenotype
MIM: 139150
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RASA1 PubMed Links

Gene Details

Gene
RASA1
Gene Name
RAS p21 protein activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002890.2 813 Silent Mutation GAC,GAG D54E NP_002881.1
NM_022650.2 813 Intron NP_072179.1
XM_011543525.1 813 Missense Mutation GAC,GAG D54E XP_011541827.1
XM_011543527.2 813 Missense Mutation GAC,GAG D54E XP_011541829.1

View Full Product Details