Product Details

SNP ID
rs201650828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29086937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTCTGTAGCACCTAGGGCCAG[C/G]AAGATGATGAGGTGGGCCACACAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2B3 PubMed Links

Gene Details

Gene
OR2B3
Gene Name
olfactory receptor family 2 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005226.2 377 Silent Mutation TTC,TTG F104L NP_001005226.1

View Full Product Details