Product Details

SNP ID
rs201613063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31971338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCACCCCTCCAACCGGCCTCGG[A/T]GTTCCAGGAGCCAGCACAGCAGGTG
Phenotype
MIM: 605996 MIM: 600478 MIM: 604977
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DXO PubMed Links

Gene Details

Gene
DXO
Gene Name
decapping exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005510.3 671 Missense Mutation CAC,CTC H113L NP_005501.2
XM_006715005.3 671 Missense Mutation CAC,CTC H113L XP_006715068.1
XM_017010329.1 671 UTR 5 XP_016865818.1
Gene
SKIV2L
Gene Name
Ski2 like RNA helicase
There are no transcripts associated with this gene.

Gene
STK19
Gene Name
serine/threonine kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004197.1 671 Intron NP_004188.1
NM_032454.1 671 Intron NP_115830.1

View Full Product Details