Product Details

SNP ID
rs201893337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55174692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTAAACCCCACCGACTATGA[A/C]GACGAGGAATTCCTGCGGTACCTGT
Phenotype
MIM: 602393
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HCRTR2 PubMed Links

Gene Details

Gene
HCRTR2
Gene Name
hypocretin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001526.3 420 Missense Mutation GAA,GAC E35D NP_001517.2
XM_017010798.1 420 Missense Mutation GAA,GAC E35D XP_016866287.1

View Full Product Details