Product Details
- SNP ID
-
rs201991278
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:56463061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTACAATACCTCTGCAGATGGCAC[A/G]GGAAAGTCTGAGCACTCTGCTGCTT
- Phenotype
-
MIM: 113810
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DST
PubMed Links
Gene Details
- Gene
- DST
- Gene Name
- dystonin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144769.2 |
16839 |
Silent Mutation |
CCC,CCT |
P5542P |
NP_001138241.1 |
NM_001144770.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5404P |
NP_001138242.1 |
NM_001723.5 |
16839 |
Intron |
|
|
NP_001714.1 |
NM_015548.4 |
16839 |
Silent Mutation |
CCC,CCT |
P5038P |
NP_056363.2 |
NM_183380.3 |
16839 |
Silent Mutation |
CCC,CCT |
P5364P |
NP_899236.1 |
XM_005249310.3 |
16839 |
Silent Mutation |
CCC,CCT |
P7685P |
XP_005249367.1 |
XM_005249315.3 |
16839 |
Silent Mutation |
CCC,CCT |
P7652P |
XP_005249372.1 |
XM_005249316.2 |
16839 |
Silent Mutation |
CCC,CCT |
P7590P |
XP_005249373.1 |
XM_005249318.2 |
16839 |
Silent Mutation |
CCC,CCT |
P7547P |
XP_005249375.1 |
XM_005249319.2 |
16839 |
Silent Mutation |
CCC,CCT |
P7514P |
XP_005249376.1 |
XM_005249320.4 |
16839 |
Silent Mutation |
CCC,CCT |
P7507P |
XP_005249377.1 |
XM_005249322.4 |
16839 |
Silent Mutation |
CCC,CCT |
P7500P |
XP_005249379.1 |
XM_005249323.3 |
16839 |
Silent Mutation |
CCC,CCT |
P7474P |
XP_005249380.1 |
XM_005249324.2 |
16839 |
Silent Mutation |
CCC,CCT |
P7148P |
XP_005249381.1 |
XM_011514824.2 |
16839 |
Silent Mutation |
CCC,CCT |
P7661P |
XP_011513126.1 |
XM_011514825.2 |
16839 |
Silent Mutation |
CCC,CCT |
P5551P |
XP_011513127.1 |
XM_011514826.2 |
16839 |
Intron |
|
|
XP_011513128.1 |
XM_017011205.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7694P |
XP_016866694.1 |
XM_017011206.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7688P |
XP_016866695.1 |
XM_017011207.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7673P |
XP_016866696.1 |
XM_017011208.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7670P |
XP_016866697.1 |
XM_017011209.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7664P |
XP_016866698.1 |
XM_017011210.1 |
16839 |
Intron |
|
|
XP_016866699.1 |
XM_017011211.1 |
16839 |
Intron |
|
|
XP_016866700.1 |
XM_017011212.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7616P |
XP_016866701.1 |
XM_017011213.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7585P |
XP_016866702.1 |
XM_017011214.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7561P |
XP_016866703.1 |
XM_017011215.1 |
16839 |
Intron |
|
|
XP_016866704.1 |
XM_017011216.1 |
16839 |
Intron |
|
|
XP_016866705.1 |
XM_017011217.1 |
16839 |
Silent Mutation |
CCC,CCT |
P7486P |
XP_016866706.1 |
XM_017011218.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5883P |
XP_016866707.1 |
XM_017011219.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5608P |
XP_016866708.1 |
XM_017011220.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5566P |
XP_016866709.1 |
XM_017011221.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5530P |
XP_016866710.1 |
XM_017011222.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5504P |
XP_016866711.1 |
XM_017011223.1 |
16839 |
Silent Mutation |
CCC,CCT |
P5499P |
XP_016866712.1 |
XM_017011224.1 |
16839 |
Intron |
|
|
XP_016866713.1 |
XM_017011225.1 |
16839 |
Intron |
|
|
XP_016866714.1 |
View Full Product Details