Product Details

SNP ID
rs202246026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55174745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAATACCTGCACCCGAAAGAAT[A/G]TGAGTGGGTCCTGATCGCCGGGTAC
Phenotype
MIM: 602393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCRTR2 PubMed Links

Gene Details

Gene
HCRTR2
Gene Name
hypocretin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001526.3 473 Missense Mutation TAT,TGT Y53C NP_001517.2
XM_017010798.1 473 Missense Mutation TAT,TGT Y53C XP_016866287.1

View Full Product Details