Product Details
- SNP ID
-
rs200880341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:152122532 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGCTGTAGTCTTCCTCTGACAT[C/T]GGTACAAGGCAGGCAAGCCCGATGA
- Phenotype
-
MIM: 608441
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYNE1
PubMed Links
Gene Details
- Gene
- SYNE1
- Gene Name
- spectrin repeat containing nuclear envelope protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033071.3 |
26758 |
Silent Mutation |
CCA,CCG |
P8718P |
NP_149062.1 |
NM_182961.3 |
26758 |
Silent Mutation |
CCA,CCG |
P8766P |
NP_892006.3 |
XM_006715407.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8815P |
XP_006715470.1 |
XM_006715408.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8811P |
XP_006715471.1 |
XM_006715409.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8808P |
XP_006715472.1 |
XM_006715410.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8801P |
XP_006715473.1 |
XM_006715411.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8798P |
XP_006715474.1 |
XM_006715412.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8796P |
XP_006715475.1 |
XM_006715413.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8792P |
XP_006715476.1 |
XM_006715414.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8791P |
XP_006715477.1 |
XM_006715415.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8778P |
XP_006715478.1 |
XM_006715416.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8773P |
XP_006715479.1 |
XM_006715417.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8768P |
XP_006715480.1 |
XM_006715420.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8764P |
XP_006715483.1 |
XM_006715421.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8763P |
XP_006715484.1 |
XM_006715422.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8762P |
XP_006715485.1 |
XM_006715423.2 |
26758 |
UTR 3 |
|
|
XP_006715486.1 |
XM_006715424.2 |
26758 |
UTR 3 |
|
|
XP_006715487.1 |
XM_006715425.2 |
26758 |
UTR 3 |
|
|
XP_006715488.1 |
XM_011535641.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8814P |
XP_011533943.1 |
XM_011535642.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8810P |
XP_011533944.1 |
XM_011535643.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8760P |
XP_011533945.1 |
XM_011535644.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8240P |
XP_011533946.1 |
XM_011535645.2 |
26758 |
Silent Mutation |
CCA,CCG |
P8071P |
XP_011533947.1 |
XM_017010608.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8815P |
XP_016866097.1 |
XM_017010609.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8815P |
XP_016866098.1 |
XM_017010610.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8808P |
XP_016866099.1 |
XM_017010611.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8806P |
XP_016866100.1 |
XM_017010612.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8789P |
XP_016866101.1 |
XM_017010613.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8777P |
XP_016866102.1 |
XM_017010614.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8763P |
XP_016866103.1 |
XM_017010615.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8726P |
XP_016866104.1 |
XM_017010616.1 |
26758 |
UTR 3 |
|
|
XP_016866105.1 |
XM_017010617.1 |
26758 |
UTR 3 |
|
|
XP_016866106.1 |
XM_017010618.1 |
26758 |
UTR 3 |
|
|
XP_016866107.1 |
XM_017010619.1 |
26758 |
Silent Mutation |
CCA,CCG |
P8240P |
XP_016866108.1 |
View Full Product Details