Product Details

SNP ID
rs200484251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149661861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCGGATAATTATATGGGTAGCC[A/G]TTGTCATCAAAAAACCTTCGGAAGG
Phenotype
MIM: 603473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LATS1 PubMed Links

Gene Details

Gene
LATS1
Gene Name
large tumor suppressor kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270519.1 3670 Intron NP_001257448.1
NM_004690.3 3670 Silent Mutation AAC,AAT N1087N NP_004681.1
XM_006715603.3 3670 Intron XP_006715666.1
XM_011536252.2 3670 Intron XP_011534554.1
XM_017011474.1 3670 Silent Mutation AAC,AAT N1120N XP_016866963.1
XM_017011475.1 3670 Silent Mutation AAC,AAT N1120N XP_016866964.1
XM_017011476.1 3670 Silent Mutation AAC,AAT N1015N XP_016866965.1
XM_017011477.1 3670 Intron XP_016866966.1
XM_017011478.1 3670 Silent Mutation AAC,AAT N982N XP_016866967.1
XM_017011479.1 3670 UTR 3 XP_016866968.1
XM_017011480.1 3670 Silent Mutation AAC,AAT N840N XP_016866969.1
XM_017011481.1 3670 Silent Mutation AAC,AAT N807N XP_016866970.1
XM_017011482.1 3670 Intron XP_016866971.1

View Full Product Details