Product Details

SNP ID
rs200684519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31829855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGGAGGGTCTGGGTCAGGCCC[C/T]ACCATTGAGGAGGTGGATTAGGGGC
Phenotype
MIM: 605447 MIM: 603012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C6orf48 PubMed Links

Gene Details

Gene
C6orf48
Gene Name
chromosome 6 open reading frame 48
There are no transcripts associated with this gene.

Gene
HSPA1B
Gene Name
heat shock protein family A (Hsp70) member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005346.4 2121 Silent Mutation CCC,CCT P635P NP_005337.2
Gene
SNORD48
Gene Name
small nucleolar RNA, C/D box 48
There are no transcripts associated with this gene.

Gene
SNORD52
Gene Name
small nucleolar RNA, C/D box 52
There are no transcripts associated with this gene.

View Full Product Details