Product Details

SNP ID
rs200369502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29673240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAATCCTCAAAGTACACTCCT[C/G]GTTTCCATCCACTGGCTCCTGGTTT
Phenotype
MIM: 159465 MIM: 612192
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 837 Intron NP_001008229.1
NM_001008229.2 837 Intron NP_001008230.1
NM_001170418.1 837 Intron NP_001163889.1
NM_002433.4 837 Intron NP_002424.3
NM_206809.3 837 Intron NP_996532.2
NM_206810.3 837 Intron NP_996533.2
NM_206811.3 837 Intron NP_996534.2
NM_206812.3 837 Intron NP_996535.2
NM_206814.5 837 Intron NP_996537.3
XM_005249131.3 837 Intron XP_005249188.1
Gene
ZFP57
Gene Name
ZFP57 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109809.2 837 Missense Mutation CAG,GAG Q291E NP_001103279.2
XM_006715087.3 837 Missense Mutation CAG,GAG Q219E XP_006715150.1
XM_011514570.2 837 Missense Mutation CAG,GAG Q291E XP_011512872.1

View Full Product Details