Product Details

SNP ID
rs200785675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87284625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTTTCTCTCTACCCTCATGATA[A/G]GCTACATGTAAAACCACCAGAAGTG
Phenotype
MIM: 611921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJB7 PubMed Links

Gene Details

Gene
GJB7
Gene Name
gap junction protein beta 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198568.2 634 Silent Mutation GCC,GCT A96A NP_940970.1
XM_005248716.2 634 Silent Mutation GCC,GCT A96A XP_005248773.1

View Full Product Details