Product Details

SNP ID
rs200591747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127577272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTTCCAATCTCTCAGAGACAGA[A/G]CCCCCTCTGTGGAAGGAGAGTCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf58 PubMed Links

Gene Details

Gene
C6orf58
Gene Name
chromosome 6 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010905.2 99 Silent Mutation GAA,GAG E29E NP_001010905.1

View Full Product Details