Product Details

SNP ID
rs201383117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:11714491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTGTGCAATTACTTCCTCTTC[C/T]TCCGTGGCTGCCTCATGTCACCTGT
Phenotype
MIM: 614348
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADTRP PubMed Links

Gene Details

Gene
ADTRP
Gene Name
androgen dependent TFPI regulating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143948.1 1022 Missense Mutation AAG,AGG K245R NP_001137420.1
NM_032744.3 1022 Missense Mutation AAG,AGG K227R NP_116133.1
XM_005249454.3 1022 Missense Mutation AAG,AGG K227R XP_005249511.1
XM_011514956.1 1022 UTR 3 XP_011513258.1
XM_011514958.2 1022 Intron XP_011513260.1
XM_011514959.1 1022 Intron XP_011513261.1
XM_017011377.1 1022 UTR 3 XP_016866866.1
Gene
LOC340184
Gene Name
uncharacterized LOC340184
There are no transcripts associated with this gene.

View Full Product Details