Product Details

SNP ID
rs201026832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116647570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCAAATAAGCGAGGGTGTGTAC[A/C]CAAAGGACCAGTTGATTTGTGAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZUFSP PubMed Links

Gene Details

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 2010 Missense Mutation GGT,TGT G453C NP_659499.2
XM_011535565.2 2010 Missense Mutation GGT,TGT G453C XP_011533867.1
XM_011535567.1 2010 Missense Mutation GGT,TGT G285C XP_011533869.1
XM_017010393.1 2010 Missense Mutation GGT,TGT G453C XP_016865882.1
XM_017010394.1 2010 Missense Mutation GGT,TGT G257C XP_016865883.1
XM_017010395.1 2010 Missense Mutation GGT,TGT G229C XP_016865884.1
XM_017010396.1 2010 Missense Mutation GGT,TGT G229C XP_016865885.1

View Full Product Details