Product Details

SNP ID
rs201482490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:50715229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCCTTTAACTTACTCCACCAC[A/C]GGCACCGAGTTTGCGTCCCCCTACT
Phenotype
MIM: 610161
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TFAP2D PubMed Links

Gene Details

Gene
TFAP2D
Gene Name
transcription factor AP-2 delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172238.3 665 Silent Mutation ACA,ACC T51T NP_758438.2

View Full Product Details