Product Details

SNP ID
rs200455262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28085685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGACCTCGCGAGGCACTGAGC[C/T]GCCTCCGGGAGCTCTGCTGTCAGTG
Phenotype
MIM: 600834
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF165 PubMed Links

Gene Details

Gene
ZNF165
Gene Name
zinc finger protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003447.3 237 Missense Mutation CGC,TGC R69C NP_003438.1
XM_017011258.1 237 Missense Mutation CGC,TGC R69C XP_016866747.1
XM_017011259.1 237 Missense Mutation CGC,TGC R69C XP_016866748.1
XM_017011260.1 237 Missense Mutation CGC,TGC R69C XP_016866749.1
XM_017011261.1 237 Intron XP_016866750.1
XM_017011262.1 237 Missense Mutation CGC,TGC R69C XP_016866751.1
Gene
ZSCAN12P1
Gene Name
zinc finger and SCAN domain containing 12 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details