Product Details

SNP ID
rs199612610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27452068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCATTCAAAGGGCTTTTCTCT[C/T]GTGTGAATTCTCCGATGTTGAGTAA
Phenotype
MIM: 602277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 1776 Silent Mutation ACA,ACG T497T NP_001305820.1
NM_001318892.1 1776 Silent Mutation ACA,ACG T497T NP_001305821.1
NM_001318893.1 1776 Silent Mutation ACA,ACG T421T NP_001305822.1
NM_007149.2 1776 Silent Mutation ACA,ACG T497T NP_009080.2
XM_011514861.2 1776 Silent Mutation ACA,ACG T421T XP_011513163.1

View Full Product Details