Product Details
- SNP ID
-
rs199534441
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30914940 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCGTTTCTACACAGTCGGAGCCCC[A/G]TGGATCTCCCATCTCCCGGAGGAAC
- Phenotype
-
MIM: 601760
MIM: 612802
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTF2H4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6926224] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTF2H4
- Gene Name
- general transcription factor IIH subunit 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001517.4 |
268 |
Intron |
|
|
NP_001508.1 |
- Gene
- VARS2
- Gene Name
- valyl-tRNA synthetase 2, mitochondrial
View Full Product Details