Product Details

SNP ID
rs199732981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116496965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGTGCTAGTCTTCATTTTTTCC[C/T]TCTATATTTTTTCTCGTCTCGCTTT
Phenotype
MIM: 614137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC3L PubMed Links

Gene Details

Gene
TRAPPC3L
Gene Name
trafficking protein particle complex 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139444.2 631 Missense Mutation AGG,GGG R179G NP_001132916.1

View Full Product Details