Product Details
- SNP ID
-
rs199656220
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:17764185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTCTCTAACTTGGAATGCAGCTG[A/G]CTGGGGTGGTGGAGCCCATCGGAGA
- Phenotype
-
MIM: 605433
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIF13A
PubMed Links
Gene Details
- Gene
- KIF13A
- Gene Name
- kinesin family member 13A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105566.2 |
5498 |
Silent Mutation |
AGC,AGT |
S1746S |
NP_001099036.1 |
NM_001105567.2 |
5498 |
Silent Mutation |
AGC,AGT |
S1733S |
NP_001099037.1 |
NM_001105568.2 |
5498 |
Silent Mutation |
AGC,AGT |
S1733S |
NP_001099038.1 |
NM_001243423.1 |
5498 |
Intron |
|
|
NP_001230352.1 |
NM_022113.5 |
5498 |
Silent Mutation |
AGC,AGT |
S1781S |
NP_071396.4 |
XM_011514806.2 |
5498 |
Intron |
|
|
XP_011513108.1 |
XM_017011188.1 |
5498 |
Silent Mutation |
AGC,AGT |
S1781S |
XP_016866677.1 |
XM_017011189.1 |
5498 |
Silent Mutation |
AGC,AGT |
S1768S |
XP_016866678.1 |
XM_017011190.1 |
5498 |
Silent Mutation |
AGC,AGT |
S1760S |
XP_016866679.1 |
XM_017011191.1 |
5498 |
Silent Mutation |
AGC,AGT |
S1747S |
XP_016866680.1 |
XM_017011192.1 |
5498 |
Silent Mutation |
AGC,AGT |
S1746S |
XP_016866681.1 |
XM_017011193.1 |
5498 |
Silent Mutation |
AGC,AGT |
S1670S |
XP_016866682.1 |
View Full Product Details