Product Details
- SNP ID
-
rs199974004
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:46688821 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTCCCGGTTCTCTCGCGAGTCCC[A/G]CTCAAGCAAAAGCAGCCTGGTCTGG
- Phenotype
-
MIM: 611200
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101926934
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41273658] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101926934
- Gene Name
- uncharacterized LOC101926934
There are no transcripts associated with this gene.
- Gene
- TDRD6
- Gene Name
- tudor domain containing 6
View Full Product Details