Product Details
- SNP ID
-
hCV191714014
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:107068932 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTACTGCAGCCGGAGGTTCTCGGC[C/G]GTGAAGAGTTCGGGAAACAGGCGGA
- Phenotype
-
MIM: 616374
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BEND3
PubMed Links
Gene Details
- Gene
- BEND3
- Gene Name
- BEN domain containing 3
There are no transcripts associated with this gene.
View Full Product Details