Product Details

SNP ID
rs200076497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTCATTACAGTTATAAGGTTTC[C/T]CTCCAGTATGAGTATTCTGATGTTC
Phenotype
MIM: 602277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 2369 Missense Mutation GAG,GGG E695G NP_001305820.1
NM_001318892.1 2369 Missense Mutation GAG,GGG E695G NP_001305821.1
NM_001318893.1 2369 Missense Mutation GAG,GGG E619G NP_001305822.1
NM_007149.2 2369 Missense Mutation GAG,GGG E695G NP_009080.2
XM_011514861.2 2369 Missense Mutation GAG,GGG E619G XP_011513163.1

View Full Product Details